Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHCBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324318.1 | NP_001311247.1 | 9 | 1211 | ||
NM_001324319.1 | NP_001311248.1 | 8 | 1091 | |||
NM_024745.4 | NP_079021.3 | 9 | 1322 | |||
NR_136738.1 | 9 | 1325 | ||||
XM_011523335.2 | XP_011521637.1 | 9 | 4856 | |||
GenBank mRNA | AK025662.1 | - | 5 | 628 | ||
AK055931.1 | - | 9 | 1293 | |||
AK225315.1 | - | 5 | 628 | |||
AK292164.1 | - | 9 | 1289 | |||
AK315706.1 | - | 9 | 1291 | |||
BC000960.1 | - | 4 | 406 | |||
BC030699.1 | - | 9 | 1297 | 4829528 |