Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145348.1 | NP_001138820.1 | 8 | 1411 | ||
NM_024773.2 | NP_079049.2 | 8 | 1361 | |||
XM_017023676.1 | XP_016879165.1 | 7 | 1268 | |||
XM_017023677.1 | XP_016879166.1 | 9 | 1446 | |||
XM_017023678.1 | XP_016879167.1 | 9 | 1442 | |||
XM_017023679.1 | XP_016879168.1 | 9 | 1442 | |||
XM_017023680.1 | XP_016879169.1 | 6 | 953 | |||
GenBank mRNA | AK023860.1 | - | 8 | 1352 | ||
AK225066.1 | - | 8 | 1318 | |||
AK297166.1 | - | 8 | 1411 | |||
AK298410.1 | - | 5 | 481 | |||
AK308499.1 | - | 6 | 611 | |||
AY345239.1 | - | 2 | 591 | |||
BC027911.1 | - | 8 | 1340 | 5207043 | ||
KU178785.1 | - | 7 | 1179 | |||
KU178786.1 | - | 6 | 1086 | |||
KU178787.1 | - | 4 | 834 |