Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024789.3 | NP_079065.2 | 8 | 1242 | ||
XM_006717973.3 | XP_006718036.1 | 10 | 1445 | |||
XM_006717974.3 | XP_006718037.1 | 7 | 1181 | |||
XM_011540160.2 | XP_011538462.1 | 9 | 1865 | |||
XM_011540163.2 | XP_011538465.1 | 9 | 1355 | |||
XM_011540167.2 | XP_011538469.1 | 6 | 668 | |||
XM_011540169.2 | XP_011538471.1 | 7 | 856 | |||
XM_017016654.1 | XP_016872143.1 | 9 | 1399 | |||
XM_017016655.1 | XP_016872144.1 | 8 | 1443 | |||
XM_017016656.1 | XP_016872145.1 | 9 | 1505 | |||
XM_017016657.1 | XP_016872146.1 | 9 | 1865 | |||
XM_017016658.1 | XP_016872147.1 | 8 | 1309 | |||
XM_017016659.1 | XP_016872148.1 | 8 | 1412 | |||
GenBank mRNA | AK026182.1 | - | 4 | 448 | ||
AK296583.1 | - | 8 | 1103 | |||
AK301612.1 | - | 7 | 799 | |||
BC057787.1 | - | 4 | 616 | |||
BC067530.1 | - | 3 | 230 | |||
BC067531.1 | - | 3 | 230 | |||
BC113589.1 | - | 6 | 1057 | 8322641 |