Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024789.3 | NP_079065.2 | 5 | 606 | ||
XM_006717973.3 | XP_006718036.1 | 7 | 809 | |||
XM_006717974.3 | XP_006718037.1 | 4 | 545 | |||
XM_011540160.2 | XP_011538462.1 | 6 | 1229 | |||
XM_011540163.2 | XP_011538465.1 | 6 | 719 | |||
XM_017016654.1 | XP_016872143.1 | 6 | 763 | |||
XM_017016655.1 | XP_016872144.1 | 5 | 807 | |||
XM_017016656.1 | XP_016872145.1 | 6 | 869 | |||
XM_017016657.1 | XP_016872146.1 | 6 | 1229 | |||
XM_017016658.1 | XP_016872147.1 | 5 | 673 | |||
XM_017016660.1 | XP_016872149.1 | 6 | 1230 | |||
XM_017016661.1 | XP_016872150.1 | 7 | 810 | |||
XM_017016662.1 | XP_016872151.1 | 6 | 764 | |||
GenBank mRNA | BC113589.1 | - | 3 | 421 | 8322641 | |
BG830198.1 | - | 5 | 607 |