Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301060.1 | NP_001287989.1 | 7, 8 | 859 | ||
NM_024848.2 | NP_079124.1 | 7, 8 | 859 | |||
XM_005244798.2 | XP_005244855.1 | 6, 7 | 723 | |||
XM_011542169.1 | XP_011540471.1 | 7, 8 | 859 | |||
XM_011542170.2 | XP_011540472.1 | 7, 8 | 859 | |||
XM_011542172.2 | XP_011540474.1 | 6, 7 | 723 | |||
XM_011542173.2 | XP_011540475.1 | 7, 8 | 859 | |||
XM_011542174.2 | XP_011540476.1 | 7, 8 | 859 | |||
XM_011542176.1 | XP_011540478.1 | 7, 8 | 859 | |||
XM_011542177.1 | XP_011540479.1 | 7, 8 | 859 | |||
XM_017002365.1 | XP_016857854.1 | 7, 8 | 859 | |||
GenBank mRNA | AK024003.1 | - | 7, 8 | 662 | ||
AK310323.1 | - | 6, 7 | 723 | |||
BC021704.2 | - | 7, 8 | 815 | 4825983 |