Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093755.1 | NP_001087224.1 | 6, 7 | 841 | ||
NM_001093756.1 | NP_001087225.1 | 6, 7 | 841 | |||
NM_001243737.1 | NP_001230666.1 | 6, 7 | 841 | |||
NM_024941.3 | NP_079217.2 | 6, 7 | 841 | |||
XM_011543652.2 | XP_011541954.1 | 5, 6 | 442 | |||
XM_017009890.1 | XP_016865379.1 | 5, 6 | 710 | |||
GenBank mRNA | AK023673.1 | - | 6, 7 | 528 | ||
BC117128.1 | - | 5, 6 | 478 | |||
BC142994.1 | - | 6, 7 | 528 | 8860449 | ||
BC143691.1 | - | 5, 6 | 478 | |||
BC143693.1 | - | 5, 6 | 478 | |||
BX641157.1 | - | 6, 7 | 571 | |||
DQ088673.1 | - | 4, 5 | 322 |