Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318330.1 | NP_001305259.1 | 14 | 1502 | ||
NM_024948.3 | NP_079224.1 | 15 | 1554 | |||
XM_005252600.2 | XP_005252657.1 | 14 | 1448 | |||
XM_005252602.2 | XP_005252659.1 | 13 | 1396 | |||
XM_006717508.2 | XP_006717571.1 | 14 | 1450 | |||
XM_011519693.1 | XP_011517995.1 | 15 | 1540 | |||
XM_011519694.1 | XP_011517996.1 | 16 | 1595 | |||
XM_017016672.1 | XP_016872161.1 | 17 | 1687 | |||
XM_017016673.1 | XP_016872162.1 | 15 | 1557 | |||
XM_017016674.1 | XP_016872163.1 | 13 | 1398 | |||
XM_017016675.1 | XP_016872164.1 | 13 | 1367 | |||
XM_017016676.1 | XP_016872165.1 | 12 | 1315 | |||
GenBank mRNA | AB014761.1 | - | 15 | 1320 | ||
AF063600.1 | - | 10 | 820 | |||
AF176916.1 | - | 14 | 1464 | |||
AK023459.1 | - | 15 | 1541 | |||
AL833873.1 | - | 15 | 1516 | |||
BC020605.1 | - | 14 | 1467 | |||
BC067799.1 | - | 15 | 1540 | 5278533 | ||
CR457315.1 | - | 15 | 1320 |