Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318330.1 | NP_001305259.1 | 7 | 864 | ||
NM_024948.3 | NP_079224.1 | 8 | 916 | |||
XM_005252600.2 | XP_005252657.1 | 7 | 810 | |||
XM_005252602.2 | XP_005252659.1 | 6 | 758 | |||
XM_006717508.2 | XP_006717571.1 | 8 | 893 | |||
XM_011519690.2 | XP_011517992.1 | 8 | 893 | |||
XM_011519691.2 | XP_011517993.1 | 8 | 893 | |||
XM_011519693.1 | XP_011517995.1 | 8 | 902 | |||
XM_011519694.1 | XP_011517996.1 | 9 | 957 | |||
XM_017016671.1 | XP_016872160.1 | 8 | 893 | |||
XM_017016672.1 | XP_016872161.1 | 10 | 1049 | |||
XM_017016673.1 | XP_016872162.1 | 8 | 919 | |||
XM_017016674.1 | XP_016872163.1 | 7 | 841 | |||
XM_017016675.1 | XP_016872164.1 | 7 | 810 | |||
XM_017016676.1 | XP_016872165.1 | 6 | 758 | |||
GenBank mRNA | AB014761.1 | - | 8 | 682 | ||
AF063600.1 | - | 3 | 182 | |||
AF176916.1 | - | 7 | 826 | |||
AK023459.1 | - | 8 | 903 | |||
AL833873.1 | - | 8 | 878 | |||
BC020605.1 | - | 7 | 829 | |||
BC067799.1 | - | 8 | 902 | 5278533 | ||
CR457315.1 | - | 8 | 682 |