Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024949.5 | NP_079225.5 | 11 | 1607 | ||
XM_011532269.2 | XP_011530571.1 | 11 | 2240 | |||
GenBank mRNA | AK126057.1 | - | 10 | 1272 | ||
AK127061.1 | - | 4 | 519 | |||
BC053873.1 | - | 2 | 183 | |||
BC107683.1 | - | 2 | 204 | |||
BX647378.1 | - | 9 | 1198 |