Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
siRNA ID
HSS129306
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
8
999
NM_001007471.2
NP_001007472.2
7
1272
NM_020952.4
NP_066003.3
7
924
NM_024971.5
NP_079247.5
7
924
NM_206944.3
NP_996827.3
7
924
NM_206945.3
NP_996828.3
7
924
NM_206946.3
NP_996829.3
8
999
NM_206947.3
NP_996830.3
8
999
NM_206948.2
NP_996831.1
7
924
XM_011519035.2
XP_011517337.1
8
1224
XM_011519036.2
XP_011517338.1
8
4149
XM_011519037.2
XP_011517339.1
8
4135
XM_011519038.2
XP_011517340.1
8
4132
XM_011519039.2
XP_011517341.1
8
4133
XM_011519040.2
XP_011517342.1
7
1155
XM_011519041.2
XP_011517343.1
7
4057
XM_011519042.2
XP_011517344.1
7
1155
XM_011519043.2
XP_011517345.1
7
1188
XM_011519044.2
XP_011517346.1
7
4058
XM_011519045.2
XP_011517347.1
7
1149
XM_011519046.2
XP_011517348.1
7
2852
XM_011519047.2
XP_011517349.1
7
2822
XM_017015141.1
XP_016870630.1
8
1224
XM_017015142.1
XP_016870631.1
8
1224
XM_017015143.1
XP_016870632.1
8
1224
XM_017015144.1
XP_016870633.1
8
990
XM_017015145.1
XP_016870634.1
8
1668
XM_017015146.1
XP_016870635.1
8
1066
XM_017015147.1
XP_016870636.1
8
3101
XM_017015148.1
XP_016870637.1
8
1224
XM_017015149.1
XP_016870638.1
8
1224
XM_017015150.1
XP_016870639.1
8
1224
XM_017015151.1
XP_016870640.1
7
1167
XM_017015152.1
XP_016870641.1
8
2902
XM_017015153.1
XP_016870642.1
7
1159
XM_017015154.1
XP_016870643.1
8
1370
XM_017015155.1
XP_016870644.1
7
2824
XM_017015156.1
XP_016870645.1
3
347
XM_017015157.1
XP_016870646.1
7
2822
XM_017015158.1
XP_016870647.1
7
2820
XM_017015159.1
XP_016870648.1
7
915
XM_017015160.1
XP_016870649.1
7
1005
XM_017015161.1
XP_016870650.1
8
1224
GenBank mRNA
AB099661.1
-
4
569
AB099662.1
-
5
644
AB099663.1
-
4
569
AB099664.1
-
5
644
AB099665.1
-
4
569
AF325212.1
-
7
895
AF536748.1
-
4
569
AF536749.1
-
4
569
AF536750.1
-
4
569
AF536751.1
-
4
569
AF536752.1
-
4
569
AF536753.1
-
5
644
AJ505025.1
-
7
1272
AJ505026.1
-
7
1272
AK225732.1
-
7
915
AK308682.1
-
7
1100
BC022454.2
-
7
929
BC067733.1
-
8
961
BC094699.1
-
7
918
BC121821.1
-
5
770
BC134414.1
-
5
773
BC142972.1
-
7
941
BM668925.1
-
1
379
KF987075.1
-
7
980
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-vrvhj:80/10.239.161.228:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0