Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025048.3 | NP_079324.2 | 4 | 449 | ||
NM_153840.3 | NP_722582.2 | 4 | 449 | |||
XM_011514469.1 | XP_011512771.1 | 2 | 164 | |||
XM_017010735.1 | XP_016866224.1 | 2 | 266 | |||
XM_017010736.1 | XP_016866225.1 | 2 | 182 | |||
GenBank mRNA | AB045123.1 | - | 4 | 403 | ||
AK026337.1 | - | 4 | 380 | |||
AK095348.1 | - | 2 | 266 | |||
AK225460.1 | - | 4 | 380 | |||
AL698860.1 | - | 4 | 433 | |||
BC029358.1 | - | 3 | 274 | |||
BC104214.1 | - | 3 | 298 | |||
BC104215.2 | - | 2 | 186 | |||
BC113634.1 | - | 3 | 262 | |||
BC113636.1 | - | 3 | 262 | |||
CR627234.1 | - | 3 | 286 | |||
DB224175.1 | - | 4 | 451 |