Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286496.1 | NP_001273425.1 | 3 | 783 | ||
NM_001286497.1 | NP_001273426.1 | 3 | 759 | |||
NM_001286499.1 | NP_001273428.1 | 3 | 783 | |||
NM_025049.3 | NP_079325.2 | 3 | 759 | |||
XM_011522083.2 | XP_011520385.1 | 3 | 875 | |||
XM_011522084.2 | XP_011520386.1 | 3 | 707 | |||
GenBank mRNA | AB185926.1 | - | 3 | 759 | ||
AB185927.1 | - | 3 | 759 | |||
AK026345.1 | - | 3 | 716 | |||
AY498716.1 | - | 2 | 683 | |||
BC137503.1 | - | 3 | 697 | 9021503 | ||
BC137504.1 | - | 3 | 697 | 9021504 | ||
DQ437529.1 | - | 2 | 664 | |||
EU084033.1 | - | 3 | 754 |