Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025057.2 | NP_079333.2 | 11 | 1652 | ||
XM_005268092.3 | XP_005268149.1 | 11 | 1658 | |||
XM_005268097.3 | XP_005268154.1 | 9 | 1019 | |||
XM_005268098.4 | XP_005268155.1 | 9 | 1291 | |||
XM_011537170.2 | XP_011535472.1 | 12 | 1766 | |||
XM_011537171.2 | XP_011535473.1 | 12 | 1766 | |||
XM_011537172.1 | XP_011535474.1 | 12 | 1766 | |||
XM_011537174.2 | XP_011535476.1 | 12 | 1766 | |||
XM_011537175.2 | XP_011535477.1 | 11 | 1622 | |||
XM_011537176.2 | XP_011535478.1 | 13 | 1750 | |||
XM_011537177.2 | XP_011535479.1 | 11 | 1470 | |||
XM_011537178.2 | XP_011535480.1 | 11 | 1441 | |||
XM_011537179.2 | XP_011535481.1 | 10 | 1326 | |||
XM_017021659.1 | XP_016877148.1 | 11 | 1658 | |||
XM_017021660.1 | XP_016877149.1 | 10 | 1374 | |||
XM_017021661.1 | XP_016877150.1 | 10 | 1426 | |||
XM_017021662.1 | XP_016877151.1 | 9 | 1210 | |||
XM_017021663.1 | XP_016877152.1 | 9 | 1019 | |||
GenBank mRNA | AK026842.1 | - | 4 | 685 | ||
AK097531.1 | - | 9 | 1330 | |||
AL834487.1 | - | 6 | 927 | |||
BC104979.1 | - | 4 | 683 | |||
BC104981.1 | - | 4 | 683 |