Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285387.2 | NP_001272316.1 | 2, 3 | 280 | ||
NM_001285404.1 | NP_001272333.1 | 3, 4 | 529 | |||
NM_025075.3 | NP_079351.2 | 3, 4 | 398 | |||
XM_006713339.3 | XP_006713402.1 | 3, 4 | 347 | |||
GenBank mRNA | AF104914.1 | - | 2, 3 | 246 | ||
AI391685.1 | - | 3, 4 | 378 | |||
AI784549.1 | - | 2, 3 | 278 | |||
AK027098.1 | - | 3, 4 | 308 | |||
AK225110.1 | - | 3, 4 | 286 | |||
BC020599.2 | - | 2, 3 | 246 | |||
BC065012.1 | - | 3, 4 | 268 | 6167514 | ||
BI914906.1 | - | 3, 4 | 529 | |||
CR457373.1 | - | 3, 4 | 265 | |||
HY023143.1 | - | 3, 4 | 398 |