Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025099.5 | NP_079375.3 | 6 | 984 | ||
NR_046431.1 | 6 | 879 | ||||
XM_006721577.3 | XP_006721640.1 | 6 | 968 | |||
XM_006721578.3 | XP_006721641.1 | 6 | 968 | |||
XM_011524010.2 | XP_011522312.1 | 6 | 863 | |||
XM_011524011.2 | XP_011522313.1 | 5 | 733 | |||
GenBank mRNA | AK091077.1 | - | 6 | 935 | ||
AL831955.1 | - | 6 | 826 | |||
BC111783.1 | - | 6 | 935 | 40054070 | ||
DB126489.1 | - | 2 | 183 | |||
DQ451688.1 | - | 6 | 925 | |||
FJ790670.1 | - | 6 | 925 |