Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM254 SIRNA Assays ›
Gene Symbol:
TMEM254
Gene Name:
transmembrane protein 254
Gene Aliases:
C10orf57, bA369J21.6
Chromosome Location:
Chr.10: 80078646 - 80092552 on Build GRCh38
UniGene ID:
Hs.169982
Species:
Human
siRNA ID
HSS129454
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM254
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270367.1
NP_001257296.1
2
174
NM_001270368.1
NP_001257297.1
2
174
NM_001270369.1
NP_001257298.1
2
174
NM_001270370.1
NP_001257299.1
2
153
NM_001270371.1
NP_001257300.1
3
248
NM_001270372.1
NP_001257301.1
2
153
NM_001270373.1
NP_001257302.1
2
153
NM_001270374.1
NP_001257303.1
3
555
NM_025125.3
NP_079401.2
2
153
NR_072984.1
3
170
NR_072985.1
3
177
NR_072986.1
2
75
NR_072987.1
2
75
XM_006717978.2
XP_006718041.1
3
505
XM_011540192.2
XP_011538494.1
2
265
XM_011540193.2
XP_011538495.1
3
486
XM_017016679.1
XP_016872168.1
2
265
XM_017016680.1
XP_016872169.1
3
505
GenBank mRNA
AK023325.1
-
2
129
AK222536.1
-
2
134
AK309425.1
-
3
452
AK309980.1
-
2
149
AK310160.1
-
3
135
AK311563.1
-
3
143
AL832397.1
-
2
50
AL832483.1
-
2
161
BC022252.1
-
2
129
4668550
BP204746.1
-
3
174
BP293793.1
-
2
128
BX537803.1
-
3
135
CR457310.1
-
2
99
CX756014.1
-
2
109
DA263618.1
-
2
33
DA520338.1
-
3
452
DA573956.1
-
2
134
DA880596.1
-
3
498
DA930892.1
-
2
129
DB485367.1
-
3
177
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0