Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270367.1 | NP_001257296.1 | 2 | 249 | ||
NM_001270368.1 | NP_001257297.1 | 2 | 249 | |||
NM_001270369.1 | NP_001257298.1 | 2 | 249 | |||
NM_001270370.1 | NP_001257299.1 | 2 | 228 | |||
NM_001270371.1 | NP_001257300.1 | 3 | 323 | |||
NM_001270372.1 | NP_001257301.1 | 2 | 228 | |||
NM_001270373.1 | NP_001257302.1 | 2 | 228 | |||
NM_001270374.1 | NP_001257303.1 | 3 | 630 | |||
NM_025125.3 | NP_079401.2 | 2 | 228 | |||
NR_072984.1 | 3 | 245 | ||||
NR_072985.1 | 3 | 252 | ||||
NR_072986.1 | 2 | 150 | ||||
NR_072987.1 | 2 | 150 | ||||
XM_006717978.2 | XP_006718041.1 | 3 | 580 | |||
XM_011540192.2 | XP_011538494.1 | 2 | 340 | |||
XM_011540193.2 | XP_011538495.1 | 3 | 561 | |||
XM_017016679.1 | XP_016872168.1 | 2 | 340 | |||
XM_017016680.1 | XP_016872169.1 | 3 | 580 | |||
GenBank mRNA | AK309425.1 | - | 3 | 527 | ||
AK309980.1 | - | 2 | 224 | |||
AL832397.1 | - | 2 | 125 | |||
BC022252.1 | - | 2 | 204 | 4668550 | ||
BP204746.1 | - | 3 | 249 | |||
BX537803.1 | - | 3 | 210 | |||
DA263618.1 | - | 2 | 108 | |||
DA520338.1 | - | 3 | 528 | |||
DA573956.1 | - | 2 | 209 | |||
DA880596.1 | - | 3 | 573 | |||
DA930892.1 | - | 2 | 204 | |||
DB485367.1 | - | 3 | 252 |