Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308445.1 | NP_001295374.1 | 5 | 816 | ||
NM_025146.3 | NP_079422.1 | 5 | 819 | |||
GenBank mRNA | AK022540.1 | - | 1 | 125 | ||
AK023090.1 | - | 5 | 734 | |||
AK023256.1 | - | 5 | 769 | |||
AK296456.1 | - | 5 | 514 | |||
AK307184.1 | - | 6 | 663 | |||
AK315876.1 | - | 5 | 621 | |||
BC012731.2 | - | 5 | 758 | 3619903 | ||
CR749314.1 | - | 3 | 450 |