Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025150.4 | NP_079426.2 | 7, 8 | 862 | ||
NR_073514.1 | 6, 7 | 737 | ||||
XM_006711555.1 | XP_006711618.1 | 7, 8 | 825 | |||
GenBank mRNA | AK222814.1 | - | 7, 8 | 803 | ||
AK298773.1 | - | 6, 7 | 690 | |||
BC000541.2 | - | 7, 8 | 809 | 3163506 | ||
BC007824.2 | - | 7, 8 | 750 | |||
BC009997.2 | - | 7, 8 | 782 | 4128881 |