Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040157.2 | NP_001035247.1 | 10 | 1470 | ||
NM_001145314.1 | NP_001138786.1 | 10 | 1361 | |||
XM_011532285.2 | XP_011530587.1 | 10 | 1421 | |||
XM_011532286.2 | XP_011530588.1 | 10 | 1470 | |||
XM_011532287.2 | XP_011530589.1 | 9 | 1320 | |||
XM_017008654.1 | XP_016864143.1 | 10 | 1421 | |||
XM_017008655.1 | XP_016864144.1 | 9 | 1320 | |||
XM_017008656.1 | XP_016864145.1 | 10 | 1470 | |||
XM_017008657.1 | XP_016864146.1 | 10 | 1421 | |||
XM_017008658.1 | XP_016864147.1 | 11 | 1602 | |||
XM_017008659.1 | XP_016864148.1 | 11 | 1555 | |||
XM_017008660.1 | XP_016864149.1 | 11 | 1720 | |||
GenBank mRNA | AK094961.1 | - | 10 | 1342 | ||
AY426674.1 | - | 8 | 1056 | |||
BC038667.1 | - | 10 | 1361 | 4823994 |