Gene Symbol: CLPTM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030782.4 | NP_110409.2 | 12 | 1482 | ||
XM_011514144.2 | XP_011512446.1 | 12 | 1446 | |||
GenBank mRNA | AB045223.1 | - | 12 | 1239 | ||
AK027306.1 | - | 12 | 1296 | |||
AK128462.1 | - | 3 | 2675 | |||
AL832953.1 | - | 2 | 84 | |||
BC016399.1 | - | 9 | 760 | |||
BC025305.1 | - | 12 | 1300 | 3051368 |