Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPTM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030782.4 | NP_110409.2 | 2, 3 | 486 | ||
XM_011514144.2 | XP_011512446.1 | 2, 3 | 453 | |||
GenBank mRNA | AB045223.1 | - | 2, 3 | 243 | ||
AK027306.1 | - | 2, 3 | 300 | |||
BC025305.1 | - | 2, 3 | 304 | 3051368 | ||
CD109529.1 | - | 2, 3 | 514 |