- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: CLPTM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030782.4 | NP_110409.2 | 3, 4 | 676 | ||
GenBank mRNA | AB045223.1 | - | 3, 4 | 433 | ||
AK027306.1 | - | 3, 4 | 490 | |||
BC016399.1 | - | 1, 2 | 62 | |||
BC025305.1 | - | 3, 4 | 494 | 3051368 |