Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271723.1 | NP_001258652.1 | 6 | 845 | ||
NM_030793.4 | NP_110420.3 | 6 | 845 | |||
XM_005268513.1 | XP_005268570.1 | 6 | 834 | |||
XM_006714797.1 | XP_006714860.1 | 6 | 834 | |||
XM_011537684.2 | XP_011535986.1 | 5 | 740 | |||
XM_017009899.1 | XP_016865388.1 | 6 | 823 | |||
XM_017009900.1 | XP_016865389.1 | 5 | 740 | |||
XM_017009901.1 | XP_016865390.1 | 5 | 740 | |||
XM_017009902.1 | XP_016865391.1 | 5 | 740 | |||
GenBank mRNA | AK024024.1 | - | 6 | 765 | ||
AK308414.1 | - | 6 | 793 | |||
BC033454.1 | - | 6 | 778 | 4510274 | ||
BC050424.1 | - | 5 | 750 | 6472693 |