Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030914.3 | NP_112176.1 | 4, 5 | 306 | ||
NR_049743.1 | 4, 5 | 347 | ||||
GenBank mRNA | AK001880.1 | - | 4, 5 | 255 | ||
BC003581.1 | - | 4, 5 | 264 | 3546655 | ||
BI823604.1 | - | 4, 5 | 305 | |||
D83200.1 | - | 3, 4 | 212 | |||
DA691216.1 | - | 4, 5 | 312 |