Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAB39L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079670.2 | NP_001073138.1 | 10 | 1123 | ||
NM_001287337.1 | NP_001274266.1 | 11 | 1261 | |||
NM_001287338.1 | NP_001274267.1 | 11 | 1256 | |||
NM_001287339.1 | NP_001274268.1 | 9 | 925 | |||
NM_030925.3 | NP_112187.2 | 8 | 1174 | |||
XM_011535254.2 | XP_011533556.1 | 12 | 1665 | |||
XM_011535255.2 | XP_011533557.1 | 10 | 1145 | |||
XM_011535256.2 | XP_011533558.1 | 11 | 1410 | |||
XM_017020785.1 | XP_016876274.1 | 11 | 1537 | |||
XM_017020786.1 | XP_016876275.1 | 12 | 1674 | |||
XM_017020787.1 | XP_016876276.1 | 12 | 1927 | |||
XM_017020788.1 | XP_016876277.1 | 13 | 1803 | |||
GenBank mRNA | AK022639.1 | - | 7 | 719 | ||
AK309011.1 | - | 5 | 449 | |||
AY288977.2 | - | 11 | 1188 | |||
BC010993.2 | - | 8 | 999 | 3956127 | ||
BX647518.1 | - | 10 | 1060 |