Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152310.2 | NP_689523.1 | 4 | 1033 | ||
XM_011540245.2 | XP_011538547.1 | 5 | 2023 | |||
GenBank mRNA | BC034344.1 | - | 4 | 925 | 4745607 |