Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297602.1 | NP_001284531.1 | 4 | 1215 | ||
NM_001297603.1 | NP_001284532.1 | 4 | 1290 | |||
NM_001297604.1 | NP_001284533.1 | 4 | 1290 | |||
NM_031299.5 | NP_112589.1 | 4 | 1290 | |||
NR_123727.1 | 4 | 1290 | ||||
GenBank mRNA | AK092246.1 | - | 4 | 493 | ||
AK291421.1 | - | 4 | 493 | |||
AK312843.1 | - | 4 | 510 | |||
BC002551.2 | - | 4 | 485 | 3138951 | ||
BM911472.1 | - | 4 | 400 | |||
BX099267.1 | - | 3 | 458 | |||
CD106434.1 | - | 4 | 754 | |||
DA735127.1 | - | 4 | 483 |