Gene Symbol: GLT8D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316967.1 | NP_001303896.1 | 11 | 1675 | ||
NM_031302.4 | NP_112592.1 | 12 | 1810 | |||
XM_011538793.2 | XP_011537095.1 | 12 | 2045 | |||
XM_017019999.1 | XP_016875488.1 | 13 | 2126 | |||
GenBank mRNA | AJ271711.1 | - | 9 | 1024 | ||
AK027310.1 | - | 10 | 1124 | |||
AK056056.1 | - | 11 | 1681 | |||
AY358763.1 | - | 11 | 1214 | |||
BC022343.1 | - | 11 | 1431 | 4248949 |