Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZMIZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300959.1 | NP_001287888.1 | 8 | 1134 | ||
NM_031449.3 | NP_113637.3 | 9 | 1303 | |||
NM_174929.2 | NP_777589.2 | 7 | 1164 | |||
XM_005249866.2 | XP_005249923.1 | 9 | 1327 | |||
XM_005249867.4 | XP_005249924.1 | 9 | 1329 | |||
XM_005249868.4 | XP_005249925.1 | 9 | 2347 | |||
XM_005249869.2 | XP_005249926.1 | 9 | 1327 | |||
XM_005249870.2 | XP_005249927.1 | 8 | 1249 | |||
XM_005249871.2 | XP_005249928.1 | 9 | 1231 | |||
XM_005249872.2 | XP_005249929.1 | 8 | 1249 | |||
XM_005249873.2 | XP_005249930.1 | 9 | 1231 | |||
XM_011515566.1 | XP_011513868.1 | 3 | 467 | |||
XM_017012674.1 | XP_016868163.1 | 9 | 1408 | |||
GenBank mRNA | AB067473.1 | - | 3 | 387 | ||
AF357908.1 | - | 8 | 1180 | |||
AK090415.1 | - | 9 | 1303 | |||
AK090841.1 | - | 1 | 583 | |||
AL831933.1 | - | 1 | 800 | |||
AY426594.1 | - | 7 | 1126 | |||
BC016968.1 | - | 7 | 1147 | |||
BC021924.1 | - | 3 | 235 | |||
BC110435.1 | - | 8 | 1119 | 5518312 |