Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMIZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300959.1 | NP_001287888.1 | 6 | 741 | ||
NM_031449.3 | NP_113637.3 | 6 | 832 | |||
NM_174929.2 | NP_777589.2 | 5 | 771 | |||
XM_005249866.2 | XP_005249923.1 | 6 | 856 | |||
XM_005249867.4 | XP_005249924.1 | 6 | 858 | |||
XM_005249868.4 | XP_005249925.1 | 6 | 1876 | |||
XM_005249869.2 | XP_005249926.1 | 6 | 856 | |||
XM_005249870.2 | XP_005249927.1 | 6 | 856 | |||
XM_005249871.2 | XP_005249928.1 | 6 | 760 | |||
XM_005249872.2 | XP_005249929.1 | 6 | 856 | |||
XM_005249873.2 | XP_005249930.1 | 6 | 760 | |||
XM_017012674.1 | XP_016868163.1 | 6 | 937 | |||
GenBank mRNA | AF357908.1 | - | 5 | 709 | ||
AK090415.1 | - | 6 | 832 | |||
AY426594.1 | - | 5 | 733 | |||
BC016968.1 | - | 5 | 754 | |||
BC110435.1 | - | 6 | 726 | 5518312 |