Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM107B SIRNA Assays ›
Gene Symbol:
FAM107B
Gene Name:
family with sequence similarity 107 member B
Gene Aliases:
C10orf45
Chromosome Location:
Chr.10: 14518557 - 14774897 on Build GRCh38
UniGene ID:
Hs.446315
Species:
Human
siRNA ID
HSS130272
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM107B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282695.1
NP_001269624.1
4
883
NM_001282696.1
NP_001269625.1
3
359
NM_001282697.1
NP_001269626.1
2
293
NM_001282698.1
NP_001269627.1
3
607
NM_001282699.1
NP_001269628.1
2
541
NM_001282700.1
NP_001269629.1
2
259
NM_001282701.1
NP_001269630.1
2
380
NM_001282702.1
NP_001269631.1
3
446
NM_001282703.1
NP_001269632.1
2
313
NM_001320735.1
NP_001307664.1
3
325
NM_001320736.1
NP_001307665.1
3
585
NM_001320737.1
NP_001307666.1
3
379
NM_001320738.1
NP_001307667.1
3
345
NM_001320739.1
NP_001307668.1
2
345
NM_001320740.1
NP_001307669.1
2
137
NM_001320741.1
NP_001307670.1
2
238
NM_031453.3
NP_113641.2
3
817
XM_005252616.3
XP_005252673.1
3
214
XM_011519735.1
XP_011518037.1
4
331
XM_017016746.1
XP_016872235.1
3
205
XM_017016747.1
XP_016872236.1
2
182
XM_017016748.1
XP_016872237.1
2
217
XM_017016749.1
XP_016872238.1
2
279
GenBank mRNA
AK074275.1
-
1
76
AK127413.1
-
3
817
AK289959.1
-
3
607
AL136885.1
-
2
303
BC004872.2
-
1
87
BC064407.1
-
2
263
BC072452.1
-
3
359
BC144707.1
-
4
691
BF979432.1
-
2
353
CR533466.1
-
1
58
DA116139.1
-
2
137
DA269906.1
-
2
163
DA305421.1
-
3
256
DA340804.1
-
3
306
DA390740.1
-
2
300
DA409460.1
-
2
319
DA412617.1
-
3
353
DA543225.1
-
2
142
DA813465.1
-
2
166
DA922675.1
-
2
238
DA954468.1
-
2
259
DA962195.1
-
2
167
DB194372.1
-
2
279
DB198416.1
-
3
291
DB240036.1
-
3
336
Back To Top
More Information
Gene Ontology Categories:
Function(s)
sensory perception of sound
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-nglqg:80/10.239.166.193:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0