Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258426.1 | NP_001245355.1 | 17 | 2371 | ||
NM_001258427.1 | NP_001245356.1 | 16 | 2224 | |||
NM_001258428.1 | NP_001245357.1 | 17 | 2485 | |||
NM_031467.2 | NP_113655.2 | 17 | 2413 | |||
XM_005268521.3 | XP_005268578.2 | 17 | 2970 | |||
XM_011537694.2 | XP_011535996.1 | 17 | 3042 | |||
XM_017009933.1 | XP_016865422.1 | 17 | 3042 | |||
XM_017009934.1 | XP_016865423.1 | 17 | 3033 | |||
XM_017009935.1 | XP_016865424.1 | 17 | 3009 | |||
XM_017009936.1 | XP_016865425.1 | 17 | 2970 | |||
XM_017009937.1 | XP_016865426.1 | 16 | 2928 | |||
XM_017009938.1 | XP_016865427.1 | 17 | 2877 | |||
XM_017009939.1 | XP_016865428.1 | 17 | 2928 | |||
XM_017009940.1 | XP_016865429.1 | 16 | 2853 | |||
XM_017009941.1 | XP_016865430.1 | 16 | 2844 | |||
XM_017009942.1 | XP_016865431.1 | 15 | 2733 | |||
XM_017009943.1 | XP_016865432.1 | 14 | 2044 | |||
GenBank mRNA | AB032762.1 | - | 17 | 2372 | ||
AF313465.1 | - | 17 | 2472 | |||
AF332961.1 | - | 17 | 2413 | |||
AF336237.1 | - | 17 | 2363 | |||
BC136262.1 | - | 16 | 2215 | 9020249 | ||
BC143602.1 | - | 16 | 2215 | 9052115 |