Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017440.2 | NP_001017440.1 | 2 | 577 | ||
NM_031468.3 | NP_113656.2 | 3 | 567 | |||
XM_011516594.2 | XP_011514896.1 | 2 | 419 | |||
XM_011516595.1 | XP_011514897.1 | 2 | 167 | |||
XM_011516596.2 | XP_011514898.1 | 3 | 1062 | |||
XM_011516597.2 | XP_011514899.1 | 2 | 995 | |||
XM_011516599.1 | XP_011514901.1 | 2 | 366 | |||
XM_017012675.1 | XP_016868164.1 | 4 | 922 | |||
XM_017012676.1 | XP_016868165.1 | 3 | 704 | |||
XM_017012677.1 | XP_016868166.1 | 2 | 217 | |||
XM_017012678.1 | XP_016868167.1 | 3 | 679 | |||
XM_017012679.1 | XP_016868168.1 | 2 | 1032 | |||
XM_017012680.1 | XP_016868169.1 | 2 | 331 | |||
XM_017012681.1 | XP_016868170.1 | 2 | 194 | |||
XM_017012682.1 | XP_016868171.1 | 2 | 744 | |||
XM_017012683.1 | XP_016868172.1 | 3 | 528 | |||
GenBank mRNA | AF282250.1 | - | 3 | 298 | ||
AK123862.1 | - | 2 | 112 | |||
AK313054.1 | - | 2 | 167 | |||
AY007302.1 | - | 2 | 225 | |||
AY726595.1 | - | 1 | 53 | |||
BC020200.1 | - | 2 | 349 | 3944302 | ||
BG718538.1 | - | 3 | 572 |