Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031474.2 | NP_113662.1 | 2 | 532 | ||
XM_005253797.3 | XP_005253854.1 | 2 | 683 | |||
XM_005253798.3 | XP_005253855.1 | 2 | 683 | |||
XM_011521031.2 | XP_011519333.1 | 3 | 474 | |||
XM_017020000.1 | XP_016875489.1 | 2 | 532 | |||
XM_017020001.1 | XP_016875490.1 | 3 | 474 | |||
GenBank mRNA | AK098063.1 | - | 2 | 503 | ||
AK300944.1 | - | 3 | 468 | |||
AL136557.1 | - | 2 | 500 | |||
BC131562.1 | - | 2 | 489 | 40109200 | ||
DA391605.1 | - | 2 | 532 |