Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127174.1 | NP_001120646.1 | 4 | 463 | ||
NM_001127175.1 | NP_001120647.1 | 4 | 574 | |||
NM_001127176.1 | NP_001120648.1 | 4 | 574 | |||
NM_031939.3 | NP_114145.2 | 5 | 677 | |||
XM_005258365.3 | XP_005258422.1 | 4 | 487 | |||
XM_011526226.2 | XP_011524528.1 | 5 | 614 | |||
XM_017026035.1 | XP_016881524.1 | 4 | 454 | |||
XM_017026036.1 | XP_016881525.1 | 5 | 614 | |||
XM_017026037.1 | XP_016881526.1 | 4 | 603 | |||
GenBank mRNA | AB042647.1 | - | 5 | 541 | ||
AI829586.1 | - | 4 | 562 | |||
AK054702.1 | - | 5 | 611 | |||
AK294757.1 | - | 4 | 418 | |||
AK295666.1 | - | 4 | 575 | |||
BC029860.1 | - | 5 | 564 | 5171860 | ||
DA416200.1 | - | 4 | 429 | |||
DB473056.1 | - | 3 | 312 | |||
DC403805.1 | - | 4 | 459 |