Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM172A SIRNA Assays ›
Gene Symbol:
FAM172A
Gene Name:
family with sequence similarity 172 member A
Gene Aliases:
C5orf21
Chromosome Location:
Chr.5: 93615130 - 94111699 on Build GRCh38
UniGene ID:
Hs.600086
Species:
Human
siRNA ID
HSS130517
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM172A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163417.1
NP_001156889.1
10
1320
NM_001163418.1
NP_001156890.1
9
1128
NM_032042.5
NP_114431.2
11
1444
NR_028080.1
9
1151
XM_005272105.3
XP_005272162.1
10
1217
XM_005272106.3
XP_005272163.1
10
1191
XM_005272108.3
XP_005272165.1
10
1338
XM_005272110.3
XP_005272167.1
9
1045
XM_006714717.3
XP_006714780.1
11
1436
XM_006714718.3
XP_006714781.1
9
2050
XM_006714719.3
XP_006714782.1
9
1199
XM_011543672.2
XP_011541974.1
11
1340
XM_011543673.2
XP_011541975.1
10
1239
XM_017009952.1
XP_016865441.1
11
1732
XM_017009953.1
XP_016865442.1
11
1730
XM_017009955.1
XP_016865444.1
10
2148
XM_017009956.1
XP_016865445.1
10
1462
XM_017009957.1
XP_016865446.1
9
1416
XM_017009958.1
XP_016865447.1
9
1211
XM_017009960.1
XP_016865449.1
8
993
GenBank mRNA
AK295879.1
-
9
1077
AK296683.1
-
5
629
AK296986.1
-
10
1297
AK297471.1
-
8
913
AK309742.1
-
2
230
AK312930.1
-
11
1349
BC020584.2
-
10
1246
4398813
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0