Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032125.2 | NP_115501.2 | 3 | 350 | ||
NR_037576.1 | 4 | 503 | ||||
NR_037577.1 | 3 | 350 | ||||
NR_037580.1 | 4 | 503 | ||||
GenBank mRNA | AF251699.1 | - | 3 | 246 | ||
AK303821.1 | - | 3 | 304 | |||
AK307077.1 | - | 4 | 341 | |||
AK313110.1 | - | 3 | 332 | |||
AL136683.1 | - | 3 | 264 | |||
BC011579.1 | - | 3 | 247 | |||
BC090039.1 | - | 4 | 395 | 6181849 | ||
BP326836.1 | - | 3 | 335 | |||
BQ927627.1 | - | 4 | 308 | |||
BX394857.2 | - | 3 | 210 | |||
DC424669.1 | - | 3 | 352 |