Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032125.2 | NP_115501.2 | 4, 5 | 452 | ||
NR_037576.1 | 5, 6 | 605 | ||||
NR_037577.1 | 4, 5 | 452 | ||||
NR_037580.1 | 5, 6 | 605 | ||||
GenBank mRNA | AF251699.1 | - | 4, 5 | 348 | ||
AK095889.1 | - | 1, 2 | 584 | |||
AK123218.1 | - | 1, 2 | 1700 | |||
AK222642.1 | - | 4, 5 | 331 | |||
AK303821.1 | - | 4, 5 | 406 | |||
AK307077.1 | - | 5, 6 | 443 | |||
AK313110.1 | - | 4, 5 | 434 | |||
AL136683.1 | - | 4, 5 | 366 | |||
BC011579.1 | - | 4, 5 | 349 | |||
BC090039.1 | - | 5, 6 | 497 | 6181849 | ||
BQ927627.1 | - | 5, 6 | 410 | |||
BX394857.2 | - | 4, 5 | 312 | |||
DC424669.1 | - | 4, 5 | 454 |