Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032125.2 | NP_115501.2 | 5 | 542 | ||
NR_037576.1 | 6 | 695 | ||||
NR_037577.1 | 5 | 542 | ||||
NR_037580.1 | 6 | 695 | ||||
GenBank mRNA | AF251699.1 | - | 5 | 438 | ||
AK095889.1 | - | 2 | 674 | |||
AK123218.1 | - | 2 | 1790 | |||
AK222642.1 | - | 5 | 421 | |||
AK303821.1 | - | 5 | 496 | |||
AK307077.1 | - | 6 | 533 | |||
AK313110.1 | - | 5 | 524 | |||
AL136683.1 | - | 5 | 456 | |||
BC011579.1 | - | 5 | 439 | |||
BC090039.1 | - | 6 | 587 | 6181849 | ||
BP326836.1 | - | 5 | 527 | |||
BQ927627.1 | - | 6 | 500 | |||
BX394857.2 | - | 5 | 402 | |||
DC424669.1 | - | 5 | 544 |