Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261467.1 | NP_001248396.1 | 6 | 734 | ||
NM_032141.3 | NP_115517.1 | 7 | 828 | |||
XM_011525345.2 | XP_011523647.1 | 7 | 743 | |||
XM_017025209.1 | XP_016880698.1 | 8 | 947 | |||
XM_017025210.1 | XP_016880699.1 | 7 | 1658 | |||
XM_017025211.1 | XP_016880700.1 | 8 | 856 | |||
XM_017025212.1 | XP_016880701.1 | 7 | 993 | |||
XM_017025213.1 | XP_016880702.1 | 6 | 773 | |||
XM_017025214.1 | XP_016880703.1 | 6 | 802 | |||
XM_017025215.1 | XP_016880704.1 | 6 | 806 | |||
GenBank mRNA | AK094552.1 | - | 2172 | |||
AK297102.1 | - | 6 | 729 | |||
AK313963.1 | - | 7 | 815 | |||
AL136806.1 | - | 7 | 819 | |||
BC016313.1 | - | 8 | 842 | |||
BC040118.1 | - | 7 | 808 | 5176197 | ||
BC046116.2 | - | 7 | 830 | |||
BC062425.1 | - | 7 | 822 | |||
BC105044.1 | - | 7 | 815 | 8144047 | ||
BC105046.1 | - | 7 | 815 | 8144049 | ||
BC143539.1 | - | 6 | 721 | |||
CR533555.1 | - | 7 | 791 |