Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042403.2 | NP_001035862.1 | 3 | 1702 | ||
NM_001278393.1 | NP_001265322.1 | 3 | 1918 | |||
NM_032147.4 | NP_115523.2 | 3 | 1847 | |||
XM_005269172.1 | XP_005269229.1 | 3 | 1841 | |||
XM_005269173.1 | XP_005269230.1 | 3 | 1918 | |||
XM_005269174.1 | XP_005269231.1 | 3 | 1685 | |||
XM_011538800.2 | XP_011537102.1 | 4 | 1868 | |||
XM_011538801.1 | XP_011537103.1 | 4 | 1728 | |||
XM_011538802.1 | XP_011537104.1 | 3 | 1688 | |||
XM_011538803.1 | XP_011537105.1 | 3 | 1693 | |||
XM_011538804.2 | XP_011537106.1 | 3 | 1901 | |||
XM_011538805.1 | XP_011537107.1 | 3 | 2542 | |||
XM_017020009.1 | XP_016875498.1 | 3 | 1836 | |||
XM_017020010.1 | XP_016875499.1 | 3 | 1688 | |||
XM_017020011.1 | XP_016875500.1 | 4 | 1714 | |||
XM_017020012.1 | XP_016875501.1 | 3 | 1841 | |||
GenBank mRNA | AK027434.1 | - | 3 | 1652 | ||
AK315697.1 | - | 3 | 1850 | |||
AL136825.1 | - | 3 | 1808 | |||
BC030704.1 | - | 3 | 1683 | 4825887 |