Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286211.1 | NP_001273140.1 | 7 | 1160 | ||
NM_001286212.1 | NP_001273141.1 | 7 | 1190 | |||
NM_001286213.1 | NP_001273142.1 | 7 | 1157 | |||
NM_032256.2 | NP_115632.1 | 8 | 1290 | |||
XM_011538831.2 | XP_011537133.1 | 8 | 1299 | |||
XM_011538832.2 | XP_011537134.1 | 8 | 974 | |||
XM_011538833.2 | XP_011537135.1 | 7 | 791 | |||
XM_017020022.1 | XP_016875511.1 | 8 | 881 | |||
XM_017020023.1 | XP_016875512.1 | 9 | 1122 | |||
GenBank mRNA | AK057287.1 | - | 8 | 1009 | ||
AK097511.1 | - | 3 | 235 | |||
AL136855.1 | - | 8 | 992 | |||
BC030284.1 | - | 7 | 898 | 5240522 | ||
BC060798.1 | - | 8 | 1053 | 30348706 |