Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGIP1 SIRNA Assays ›
Gene Symbol:
SGIP1
Gene Name:
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 66533272 - 66751139 on Build GRCh38
UniGene ID:
Hs.132121
Species:
Human
siRNA ID
HSS130804
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308203.1
NP_001295132.1
18
1627
NM_032291.3
NP_115667.2
21
2404
XM_005271264.3
XP_005271321.1
21
2238
XM_005271268.3
XP_005271325.1
19
1653
XM_005271270.4
XP_005271327.1
18
1960
XM_006710961.2
XP_006711024.1
21
2244
XM_006710966.2
XP_006711029.1
20
2178
XM_006710967.2
XP_006711030.1
20
2166
XM_006710969.2
XP_006711032.1
20
2154
XM_006710971.2
XP_006711034.1
20
1743
XM_006710972.2
XP_006711035.1
20
1737
XM_006710973.2
XP_006711036.1
18
1719
XM_006710974.2
XP_006711037.1
17
1635
XM_011542291.1
XP_011540593.1
20
1900
XM_011542292.1
XP_011540594.1
19
1840
XM_011542293.1
XP_011540595.1
16
1569
XM_017002505.1
XP_016857994.1
22
2292
XM_017002506.1
XP_016857995.1
23
2415
XM_017002507.1
XP_016857996.1
22
2467
XM_017002508.1
XP_016857997.1
23
2413
XM_017002509.1
XP_016857998.1
20
2268
XM_017002510.1
XP_016857999.1
21
2391
XM_017002511.1
XP_016858000.1
22
2349
XM_017002512.1
XP_016858001.1
22
2222
XM_017002513.1
XP_016858002.1
21
2401
XM_017002514.1
XP_016858003.1
21
2220
XM_017002515.1
XP_016858004.1
19
2196
XM_017002516.1
XP_016858005.1
20
2319
XM_017002517.1
XP_016858006.1
21
2288
XM_017002518.1
XP_016858007.1
21
2276
XM_017002519.1
XP_016858008.1
21
2301
XM_017002520.1
XP_016858009.1
19
2142
XM_017002521.1
XP_016858010.1
19
2094
XM_017002522.1
XP_016858011.1
20
2216
XM_017002523.1
XP_016858012.1
20
2240
XM_017002524.1
XP_016858013.1
20
1883
XM_017002525.1
XP_016858014.1
19
1822
XM_017002526.1
XP_016858015.1
18
1701
XM_017002527.1
XP_016858016.1
19
1725
XM_017002528.1
XP_016858017.1
19
1811
XM_017002529.1
XP_016858018.1
19
1677
XM_017002530.1
XP_016858019.1
18
1653
XM_017002531.1
XP_016858020.1
17
1653
XM_017002532.1
XP_016858021.1
18
1647
XM_017002533.1
XP_016858022.1
18
1755
XM_017002534.1
XP_016858023.1
17
1629
XM_017002535.1
XP_016858024.1
18
1743
XM_017002536.1
XP_016858025.1
18
1593
XM_017002537.1
XP_016858026.1
17
1682
GenBank mRNA
AB210039.1
-
21
2186
AK090697.1
-
12
1184
AK125044.1
-
4
1171
AK298970.1
-
18
1590
AK299941.1
-
19
1664
AL136561.1
-
21
2078
BC040516.1
-
23
2232
5275011
BX640813.1
-
18
1604
CR749541.1
-
18
1815
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
positive regulation of receptor-mediated endocytosis
membrane tubulation
protein binding
phospholipid binding
microtubule binding
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-k7c2b:80/10.239.160.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0