Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032323.2 | NP_115699.1 | 3 | 1044 | ||
NR_026678.1 | 3 | 1069 | ||||
GenBank mRNA | AK056816.1 | - | 2 | 1285 | ||
AK122634.1 | - | 4 | 790 | |||
AK315767.1 | - | 3 | 1012 | |||
BC005094.1 | - | 3 | 1034 | 3955668 |