Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3MBTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007102.2 | NP_001007103.1 | 22 | 2369 | ||
NM_032438.2 | NP_115814.1 | 23 | 2444 | |||
XM_005267161.4 | XP_005267218.1 | 23 | 2620 | |||
XM_006715576.3 | XP_006715639.1 | 23 | 2549 | |||
XM_006715578.3 | XP_006715641.1 | 23 | 2428 | |||
XM_011536179.2 | XP_011534481.1 | 23 | 2427 | |||
XM_011536180.2 | XP_011534482.1 | 23 | 2446 | |||
XM_011536181.2 | XP_011534483.1 | 23 | 2381 | |||
XM_011536183.2 | XP_011534485.1 | 23 | 2977 | |||
XM_011536184.2 | XP_011534486.1 | 23 | 2381 | |||
XM_017011355.1 | XP_016866844.1 | 22 | 2460 | |||
XM_017011356.1 | XP_016866845.1 | 22 | 2352 | |||
GenBank mRNA | AB058701.1 | - | 22 | 2342 | ||
AK092756.1 | - | 13 | 1621 | |||
AK094137.1 | - | 23 | 2451 | |||
AK290219.1 | - | 22 | 2361 | |||
BC060845.1 | - | 22 | 2363 | 30342272 |