Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032451.1 | NP_115827.1 | 8 | 1288 | ||
XM_011523379.1 | XP_011521681.1 | 8 | 2054 | |||
XM_011523380.2 | XP_011521682.1 | 9 | 1310 | |||
XM_011523381.1 | XP_011521683.1 | 9 | 2134 | |||
XM_011523384.1 | XP_011521686.1 | 8 | 1312 | |||
XM_011523385.1 | XP_011521687.1 | 8 | 1312 | |||
XM_017023779.1 | XP_016879268.1 | 8 | 1300 | |||
XM_017023780.1 | XP_016879269.1 | 8 | 1465 | |||
XM_017023781.1 | XP_016879270.1 | 7 | 937 | |||
GenBank mRNA | AB058735.1 | - | 6 | 1386 | ||
AJ422077.1 | - | 8 | 1278 | |||
AK094918.1 | - | 1 | 105 | |||
AL834408.1 | - | 8 | 1082 | |||
BC111030.1 | - | 8 | 1288 | 5244492 | ||
BC139732.1 | - | 6 | 906 | 40075864 |