Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032833.4 | NP_116222.4 | 1 | 1056 | ||
XM_005245551.4 | XP_005245608.2 | 1 | 1119 | |||
GenBank mRNA | AK222877.1 | - | 1 | 1057 | ||
AK303535.1 | - | 2 | 610 | |||
AK316092.1 | - | 2 | 668 | |||
BC065280.1 | - | 1 | 1030 | 6172811 |