Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMODL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004416.2 | NP_001004416.2 | 20 | 3595 | ||
NM_001199527.1 | NP_001186456.1 | 19 | 4159 | |||
NM_001199528.2 | NP_001186457.2 | 20 | 3775 | |||
NM_173568.3 | NP_775839.3 | 19 | 3979 | |||
XM_011529797.1 | XP_011528099.1 | 17 | 3587 | |||
XM_017028506.1 | XP_016883995.1 | 19 | 3974 | |||
XM_017028507.1 | XP_016883996.1 | 19 | 6903 | |||
XM_017028508.1 | XP_016883997.1 | 9 | 1758 | |||
GenBank mRNA | AB051810.1 | - | 19 | 4159 | ||
AB051811.1 | - | 20 | 3775 | |||
AB096973.1 | - | 1 | 59 | |||
AK093654.1 | - | 5 | 837 | |||
AK124223.1 | - | 6 | 1339 | |||
AY771619.1 | - | 19 | 3979 | |||
AY771620.1 | - | 20 | 3595 |