Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173462.3 | NP_775733.3 | 6 | 449 | ||
XM_011537290.2 | XP_011535592.1 | 6 | 449 | |||
XM_011537291.2 | XP_011535593.1 | 7 | 496 | |||
XM_011537292.2 | XP_011535594.1 | 6 | 372 | |||
XM_011537293.2 | XP_011535595.1 | 5 | 368 | |||
XM_011537294.2 | XP_011535596.1 | 5 | 373 | |||
XM_011537295.2 | XP_011535597.1 | 5 | 368 | |||
XM_011537296.2 | XP_011535598.1 | 5 | 368 | |||
XM_017021749.1 | XP_016877238.1 | 5 | 368 | |||
XM_017021750.1 | XP_016877239.1 | 5 | 371 | |||
GenBank mRNA | AK293773.1 | - | 5 | 510 | ||
AK294560.1 | - | 5 | 501 | |||
BC042057.1 | - | 6 | 399 | |||
BC136623.1 | - | 5 | 367 | 9020613 | ||
BC144315.1 | - | 5 | 367 | |||
BX470414.1 | - | 2 | 108 | |||
DB088927.1 | - | 6 | 419 |
Gene Symbol: NDRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098388.1 | - | 401 |